missing translation for 'onlineSavingsMsg'
Få mere at vide

9-Chloro Quetiapine, TRC

Artikelnummer. 30353552
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353552 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30353552 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36604025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 9-Chloro Quetiapine
CAS 1371638-11-7
Molekylær formel C21H24ClN3O2S
Mængde 25 mg
Synonym 2-[2-[4-(9-Chlorodibenzo[b,f][1,4]thiazepin-11-yl)-1-piperazinyl]ethoxy]ethanol, Quetiapine Impurity L
InChI formel InChI=1S/C21H24ClN3O2S/c22-17-5-3-7-19-20(17)23-21(16-4-1-2-6-18(16)28-19)25-10-8-24(9-11-25)12-14-27-15-13-26/h1-7,26H,8-15H2
SMIL OCCOCCN1CCN(CC1)C2=Nc3c(Cl)cccc3Sc4ccccc24
IUPAC navn 2-[2-[4-(4-chlorobenzo[b][1,4]benzothiazepin-6-yl)piperazin-1-yl]ethoxy]ethanol
Molekylvægt (g/mol) 417.95
Formel vægt 417.1278
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.