Få mere at vide
9-Carboxy-10-methylacridinium Trifluoromethanesulfonic Acid Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 9-Carboxy-10-methylacridinium Chloride |
| CAS | 173920-64-4 |
| Molekylær formel | C16H12F3NO5S |
| Mængde | 5 mg |
| Synonym | Acridinium, 9-carboxy-10-methyl-, 1,1,1-trifluoromethanesulfonate (1:1), Acridinium, 9-carboxy-10-methyl-, salt with trifluoromethanesulfonic acid (1:1) (9CI), 10-methylacridin-10-ium-9-carboxylate compound with trifluoromethanesulfonic acid (1:1), 9-Carboxy-10-methylacridinium trifluoromethanesulfonic acid salt, 9-Carboxy-10-methylacridinium trifluoromethanesulfonate |
| InChI formel | InChI=1S/C15H11NO2.CHF3O3S/c1-16-12-8-4-2-6-10(12)14(15(17)18)11-7-3-5-9-13(11)16;2-1(3,4)8(5,6)7/h2-9H,1H3;(H,5,6,7) |
| SMIL | C[n+]1c2ccccc2c(C(=O)O)c3ccccc13.[O-]S(=O)(=O)C(F)(F)F |
| IUPAC navn | 10-methylacridin-10-ium-9-carboxylic acid;trifluoromethanesulfonate |
| Molekylvægt (g/mol) | 387.33 |
| Formel vægt | 387.0388 |