missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9-Bromo-10-(2-naphthyl)anthracene, 98%

Brand:  Alfa Aesar™ H64746.03

Additional Details : CAS Number : 474688-73-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402069

  • 600.58 DKK / 1g


Estimated Shipment date
from Supplier 19-11-2018
Add to basket

Specifications

Chemical Identifiers

474688-73-8
383.288
FKIFDWYMWOJKTQ-UHFFFAOYSA-N
16116263
C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br
C24H15Br
MFCD09832882
9-bromo-10-2-naphthyl anthracene, 9-bromo-10-naphthalen-2-yl anthracene, 10-bromo-9-naphthalen-2-yl anthracene, anthracene, 9-bromo-10-2-naphthalenyl, ksc235k9l, 10-2-naphthyl-9-anthryl bromide, 9-bromo-10-2-naphthyl-anthracene, 9-bromo-10-2-naphthyl anthracene, 10-bromo-9-phthalen-2-yl anthracene, 9-bromo-10-naphthyl-2-yl anthracene
9-bromo-10-naphthalen-2-ylanthracene

Specifications

9-Bromo-10-(2-naphthyl)anthracene
1g
C24H15Br
10698181
FKIFDWYMWOJKTQ-UHFFFAOYSA-N
9-bromo-10-naphthalen-2-ylanthracene
16116263
Solid
171°C to 173°C
474688-73-8
MFCD09832882
9-bromo-10-2-naphthyl anthracene, 9-bromo-10-naphthalen-2-yl anthracene, 10-bromo-9-naphthalen-2-yl anthracene, anthracene, 9-bromo-10-2-naphthalenyl, ksc235k9l, 10-2-naphthyl-9-anthryl bromide, 9-bromo-10-2-naphthyl-anthracene, 9-bromo-10-2-naphthyl anthracene, 10-bromo-9-phthalen-2-yl anthracene, 9-bromo-10-naphthyl-2-yl anthracene
C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br
383.288
383.29
98%
Videos
SDS