missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Anthracenemethanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.
Solubility
Soluble in hot methanol very faint turbidity.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.
Solubility
Soluble in hot methanol very faint turbidity.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Melting Point | 160°C to 165°C |
Quantity | 10 g |
Beilstein | 1873402 |
Solubility Information | Soluble in hot methanol very faint turbidity. |
Formula Weight | 208.26 |
Percent Purity | 97% |
Chemical Name or Material | 9-Anthracenemethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'