missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-pentylfluorene-2-boronic acid, 97%, Thermo Scientific™

Catalog Number 15441808
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
400607-33-2
C23H31BO2
350.309
MFCD22581216
VCMFLHIBNVHQOD-UHFFFAOYSA-N
9,9-di-n-pentylfluorene-2-boronic acid
22348459
(9,9-dipentylfluoren-2-yl)boronic acid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(CCCCC)CCCCC)(O)O
This item is not returnable. View return policy
400607-33-2
C23H31BO2
350.309
MFCD22581216
VCMFLHIBNVHQOD-UHFFFAOYSA-N
9,9-di-n-pentylfluorene-2-boronic acid
22348459
(9,9-dipentylfluoren-2-yl)boronic acid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(CCCCC)CCCCC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 394.12
Percent Purity 97%
Chemical Name or Material 9,9-Di-n-pentylfluorene-2-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'