Learn More
Alfa Aesar™ 9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%
Brand: Alfa Aesar™ H55817.MD
Additional Details : CAS Number : 203927-98-4 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
203927-98-4 | |
422.179 | |
QFWOJNOZWMHNTK-UHFFFAOYSA-N | |
4524285 | |
B(C1=CC2=C(C=C1)C3=C(C2(CCCCCC)CCCCCC)C=C(C=C3)B(O)O)(O)O |
C25H36B2O4 | |
MFCD03427279 | |
9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene | |
(7-borono-9,9-dihexylfluoren-2-yl)boronic acid |
Specifications
9,9-Di-n-hexylfluorene-2,7-diboronic acid | |
203927-98-4 | |
MFCD03427279 | |
QFWOJNOZWMHNTK-UHFFFAOYSA-N | |
(7-borono-9,9-dihexylfluoren-2-yl)boronic acid | |
4524285 | |
Solid |
250mg | |
C25H36B2O4 | |
9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene | |
B(C1=CC2=C(C=C1)C3=C(C2(CCCCCC)CCCCCC)C=C(C=C3)B(O)O)(O)O | |
422.179 | |
422.18 | |
97% |