missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(4-Bromophenyl)carbazole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 147°C to 151°C |
Quantity | 5 g |
Beilstein | 199590 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 322.2 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 9-(4-Bromophenyl)carbazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'