missing translation for 'onlineSavingsMsg'
Få mere at vide

9,10-Phenanthrenequinone, Sigma-Aldrich™

Artikelnummer. 15824221
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15824221 100 g
15814221 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15824221 Leverandør Sigma-Aldrich Leverandørnr. 8075640100
 Begrænset vare
Forventet forsendelse: 23-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

For synthesis

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 84-11-7
Grad Research
Molekylvægt (g/mol) 208.21

Tekniske data

Smeltepunkt 203°C to 206°C
Kogepunkt >360°C
MDL nummer MFCD00001163
Mængde 100 g
Synonym 9,10-Phenanthrenequinone, 9,10-Phenanthrenedione, 9,10-Phenanthrenequinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.