Få mere at vide
9,10-Dibromoanthracene-2-sulfonyl Chloride, TRC
Artikelnummer. 30373188
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373188 100 mg
30376130 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30373188 Leverandør Toronto Research Chemicals Leverandørnr. TRCD423500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9,10-Dibromoanthracene-2-sulfonyl Chloride
CAS 210174-74-6
Molekylær formel C14 H7 Br2 Cl O2 S
Mængde 100 mg
InChI formel InChI=1S/C14H7Br2ClO2S/c15-13-9-3-1-2-4-10(9)14(16)12-7-8(20(17,18)19)5-6-11(12)13/h1-7H
SMIL ClS(=O)(=O)c1ccc2c(Br)c3ccccc3c(Br)c2c1
IUPAC navn 9,10-dibromoanthracene-2-sulfonyl chloride
Molekylvægt (g/mol) 434.53
Formel vægt 431.8222