Få mere at vide
Tocris Bioscience™ 8MDP
Potent equilibrative nucleoside transporter (ENT) inhibitor
Brand: Tocris Bioscience™ 4588/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
8MDP is a potent equilibrative nucleoside transporter 1 (ENT1) inhibitor (IC50 = 0.43 nM). Inhibits hENT1 and hENT2 uptake of [H3]uridine in K562 cells and K15NTD cells respectively.Tekniske data
| 2,2′,2′′,2′′′-[[4,8-Bis(hexahydro-1(2H)-azocinyl)pyrimido[5,4-d]pyrimidine-2,6-diyl]dinitrilo]tetrakisethanol | |
| 29491-75-6 | |
| 10 mg |