Learn More
Moldex™ 8000 Particulate Filters
Product Code. 12302319
P3RD, Pack of 8
Click to view available options
Filter Type:
P2RD
P3RD
Unit Size:
Pack of 8
For the 8000 Series half mask, protects against harmful and carcinogenic dusts
Moldex 8000 Series P2 R D filters (8070). P2 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols based on water and oil. 10 x WEL.
Moldex 8000 Series P3 R D filters (8080). P3 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols including radioactive particulates as well as airborne biological substances and enzymes. 20 x WEL.
Specifications
Filter Type P3RD
Product Type Particulate Filter
For Use With (Equipment) For Series 8000 Respiratory Filter
Quantity 8/Pk.
Product Content Correction
Product Title
Moldex™ 8000 Particulate Filters >