Learn More
Hamilton™ 800 Series Replacement Barrel/Plunger Assembly
The reinforced plunger version of the 700 series syringes.
Brand: Hamilton™ 32134
Additional Details : Weight : 0.04700kg
Description
- Attached to the flange of the syringe is a blue syringe holder
- Syringe holder provides an area to hold the syringe that prevents heat transfer, dispense inaccuracies, and plunger blow-outs
- Provides support to small volume plungers that are prone to bending during injection
Specifications
5 μL | |
Removable |