Learn More
Hamilton™ 800 Series Replacement Barrel/Plunger Assembly
The reinforced plunger version of the 700 series syringes.
Brand:  Hamilton™ 32134
Additional Details : Weight : 0.04700kg
Product Code. 11592465
  • 951.00 DKK / Each
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
  • Attached to the flange of the syringe is a blue syringe holder
  • Syringe holder provides an area to hold the syringe that prevents heat transfer, dispense inaccuracies, and plunger blow-outs
  • Provides support to small volume plungers that are prone to bending during injection
Specifications
Specifications
5 μL
Removable