Få mere at vide
8-Oxoadenosine, TRC
Artikelnummer. 30428221
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428221 50 mg
30422484 10 mg
30421343 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428221 Leverandør Toronto Research Chemicals Leverandørnr. TRCO84726350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 29851-57-8
Molekylær formel C10H13N5O5
Molekylvægt (g/mol) 283.24
Synonym NSC 102240, NSC 90396, 7,8-Dihydro-8-oxoadenosine, 8-Hydroxyadenosine, 8-Oxo-7,8-dihydroadenosine, 8-Oxyadenosine, 6-Amino-7,9-dihydro-9-beta-D-ribofuranosyl-8H-purin-8-one
IUPAC navn (2R,3R,4S,5R)-2-(6-amino-8-hydroxypurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMIL Nc1ncnc2c1nc(O)n2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
Tekniske data
Kemisk navn eller materiale 8-Oxoadenosine
Mængde 50 mg
InChI formel InChI=1S/C10H13N5O5/c11-7-4-8(13-2-12-7)15(10(19)14-4)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H,14,19)(H2,11,12,13)/t3-,5-,6-,9-/m1/s1
Formel vægt 283.0917