Få mere at vide
8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt, Mikromol™
Artikelnummer. 17950696
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17950696 100 mg
17923586 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17950696 Leverandør Mikromol Leverandørnr. MM0599.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
CAS 64487-94-1
Analyt- eller komponentnavne 8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
Forsendelsestilstand Room Temperature
Molekylær formel C24 H27 N5 O3
Certificeringer/compliance ISO 17025
Synonym 8-[N-(2-Diphenylmethoxyethyl)-N,N-dimethylaminium]-1,3-dimethylxanthine inner salt
InChI formel InChI=1S/C24H27N5O3/c1-27-21-19(22(30)28(2)24(27)31)25-23(26-21)29(3,4)15-16-32-20(17-11-7-5-8-12-17)18-13-9-6-10-14-18/h5-14,20H,15-16H2,1-4H3
SMIL CN1C(=O)N(C)c2nc([n-]c2C1=O)[N+](C)(C)CCOC(c3ccccc3)c4ccccc4
IUPAC navn 2-benzhydryloxyethyl-(1,3-dimethyl-2,6-dioxopurin-7-id-8-yl)-dimethylazanium
Molekylvægt (g/mol) 433.5
Mængde 100 mg
Formel vægt 433.2114 g/mol