missing translation for 'onlineSavingsMsg'
Få mere at vide

8-Methoxypyrene-1,3,6-trisulfonic Acid Trisodium Salt, TRC

Artikelnummer. 30473546
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30473546 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473546 Supplier Toronto Research Chemicals Supplier No. TRCM33294050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 8-Methoxypyrene-1,3,6-trisulfonic Acid Trisodium Salt
CAS 82962-86-5
Molekylær formel C24 H38 N2 O
Mængde 50 mg
Synonym Trisodium 8-Methoxy-1,3,6-pyrenetrisulfonate, 8-Methoxypyrene-1,3,6-trisulfonic Acid Sodium Salt (1:3)
InChI formel InChI=1S/C24H38N2O/c1-27-24-14-6-12-22-20(8-5-13-23(22)24)9-7-15-25-16-18-26(19-17-25)21-10-3-2-4-11-21/h6,12,14,20-21H,2-5,7-11,13,15-19H2,1H3
SMIL COc1cccc2C(CCCN3CCN(CC3)C4CCCCC4)CCCc12
IUPAC navn 1-cyclohexyl-4-[3-(5-methoxy-1,2,3,4-tetrahydronaphthalen-1-yl)propyl]piperazine
Molekylvægt (g/mol) 370.571
Formel vægt 370.298
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.