Få mere at vide
8-Methoxy Psoralen-13CD3, TRC
Artikelnummer. 30475266
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30475266 25 mg -
30478557 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475266 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26079725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246819-63-5
Molekylær formel C1113CH5D3O4
Molekylvægt (g/mol) 220.2
Synonym 9-(Methoxy-13CD3)-7H-furo[3,2-g][1]benzopyran-7-one, 6-Hydroxy-7-(methoxy-13CD3)-5-benzofuranacrylic Acid δ-Lactone, 8-MOP-13CD3, 8-MP-13CD3, 8-(Methoxy-13CD3)-6,7-furanocoumarin, 8-(Methoxy-13CD3)[furano-3',2':6,7-coumarin], 8-(Methoxy-13CD3)psoralene, Ammodin-13CD3, Ammoidin-13CD3, Deltapsoralen-13CD3, Geroxalen-13CD3, Meladinin-13CD3, Meladinine-13CD3, Meladoxen-13CD3, Meloxine-13CD3, Methoxsalen-13CD3, Methoxsalene-13CD3, NSC 45923-13CD3, New-Meladinin-13CD3, Oxsoralen-13CD3, Oxypsoralen-13CD3, Puvalen-13CD3, Puvamet-13CD3, Uvadex-13CD3, Vitpso-13CD3, Xanthotoxin-13CD3, Xanthotoxine-13CD3
SMIL O=C(C=C1)OC2=C1C=C(C=CO3)C3=C2O[13C]([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale 8-Methoxy Psoralen-13CD3
Mængde 25 mg
InChI formel InChI=1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3