missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-4509660
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
283158-18-9
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
2723649
8-hydroxyquinoline-5-sulfonic acid;hydrate
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O
This item is not returnable. View return policy
283158-18-9
WUXYGIQVWKDVTJ-UHFFFAOYSA-N
8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
2723649
8-hydroxyquinoline-5-sulfonic acid;hydrate
C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass Bottle
Molecular Formula C9H7NO4S·H2O
Quantity 5 g
Beilstein 22,407
Merck Index 14,4844
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in most common organic solvents
Molecular Weight (g/mol) 243.24
Formula Weight 243.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 8-Hydroxyquinoline-5-sulfonic acid monohydrate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'