missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxyjulolidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
Specifications
Melting Point | 132°C to 135°C |
Quantity | 1 g |
Beilstein | 1530423 |
Synonym | 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol |
Solubility Information | Soluble in water (partly),ethanol,chloroform,methanol,and acetone. |
SMILES | C1CC2=C3C(=C(C=C2)O)CCCN3C1 |
Molecular Weight (g/mol) | 189.258 |
Formula Weight | 189.26 |
Percent Purity | 97% |
Chemical Name or Material | 8-Hydroxyjulolidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'