Få mere at vide
8-Desmethoxy-8-fluoro Moxifloxacin, TRC
Artikelnummer. 30010454
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010454 25 mg
30004784 1 mg
30008398 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010454 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44585025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 151213-15-9
Molekylær formel C20 H21 F2 N3 O3
Molekylvægt (g/mol) 389.4
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,8-difluoro-1,4-dihydro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-, 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,8-difluoro-1,4-dihydro-7-(octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl)-4-oxo-, (4aS-cis)-, 6H-Pyrrolo[3,4-b]pyridine, 3-quinolinecarboxylic acid deriv.
IUPAC navn 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6,8-difluoro-4-oxoquinoline-3-carboxylic acid
SMIL OC(=O)C1=CN(C2CC2)c3c(F)c(N4C[C@@H]5CCCN[C@@H]5C4)c(F)cc3C1=O
Tekniske data
Kemisk navn eller materiale 1-Cyclopropyl-6,8-difluoro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid
Mængde 25 mg
InChI formel InChI=1S/C20H21F2N3O3/c21-14-6-12-17(25(11-3-4-11)8-13(19(12)26)20(27)28)16(22)18(14)24-7-10-2-1-5-23-15(10)9-24/h6,8,10-11,15,23H,1-5,7,9H2,(H,27,28)/t10-,15+/m0/s1
Formel vægt 389.1551