missing translation for 'onlineSavingsMsg'
Få mere at vide

8-Demethyl Ivabradine, TRC

Artikelnummer. 30350757
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30350757 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350757 Supplier Toronto Research Chemicals Supplier No. TRCD29299025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 8-Demethyl Ivabradine
CAS 304464-97-9
Molekylær formel C26 H34 N2 O5
Mængde 25 mg
Synonym 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-8-hydroxy-7-methoxy- (9CI, ACI), 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-8-hydroxy-7-methoxy-2H-3-benzazepin-2-one (ACI), S 33174, 8-Desmethyl Ivabradine
InChI formel InChI=1S/C26H34N2O5/c1-27(16-20-10-19-13-24(32-3)25(33-4)15-21(19)20)7-5-8-28-9-6-17-12-23(31-2)22(29)11-18(17)14-26(28)30/h11-13,15,20,29H,5-10,14,16H2,1-4H3/t20-/m1/s1
SMIL COc1cc2CCN(CCCN(C)C[C@H]3Cc4cc(OC)c(OC)cc34)C(=O)Cc2cc1O
IUPAC navn 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7-hydroxy-8-methoxy-2,5-dihydro-1H-3-benzazepin-4-one
Molekylvægt (g/mol) 454.559
Formel vægt 454.247
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.