Få mere at vide
8-Dechloro-7-chloro-N-methyl Desloratadine, TRC
Artikelnummer. 30357752
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357752 50 mg
30355473 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357752 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22635050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346600-30-3
Molekylær formel C20 H21 Cl N2
Molekylvægt (g/mol) 324.847
Synonym 7-Chloro-6,11-dihydro-11-(1-methyl-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
IUPAC navn 12-chloro-2-(1-methylpiperidin-4-ylidene)-4-azatricyclo[9.4.0.03,8]pentadeca-1(11),3(8),4,6,12,14-hexaene
SMIL CN1CCC(=C2c3cccc(Cl)c3CCc4cccnc24)CC1
Tekniske data
Kemisk navn eller materiale 8-Dechloro-7-chloro-N-methyl Desloratadine
Mængde 50 mg
InChI formel InChI=1S/C20H21ClN2/c1-23-12-9-14(10-13-23)19-17-5-2-6-18(21)16(17)8-7-15-4-3-11-22-20(15)19/h2-6,11H,7-10,12-13H2,1H3
Formel vægt 324.139