Få mere at vide
8-Cyclopentyl-1,3-dimethylxanthine, TRC
Artikelnummer. 30357106
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357106 1 g
30360637 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357106 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9884901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 8-Cyclopentyl-1,3-dimethylxanthine
CAS 35873-49-5
Molekylær formel C12 H16 N4 O2
Mængde 1 g
InChI formel InChI=1S/C12H16N4O2/c1-15-10-8(11(17)16(2)12(15)18)13-9(14-10)7-5-3-4-6-7/h7H,3-6H2,1-2H3,(H,13,14)
SMIL CN1C(=O)N(C)c2nc([nH]c2C1=O)C3CCCC3
IUPAC navn 8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
Molekylvægt (g/mol) 248.28
Formel vægt 248.1273