Få mere at vide
8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]diazepin-11-one, Mikromol™
Artikelnummer. 17963686
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963686 25 mg
17920986 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17963686 Leverandør Mikromol Leverandørnr. MM0480.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]diazepin-11-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]diazepin-11-one
CAS 50892-62-1
Analyt- eller komponentnavne 8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]diazepin-11-one
Forsendelsestilstand Room Temperature
Molekylær formel C13 H9 Cl N2 O
Certificeringer/compliance ISO 17025
Synonym 8-Chloro-5,10-dihydro-11H-dibenzo[b,e][1,4]diazepin-11-one, Clozapine Imp. A (EP), Clozapine Impurity A
InChI formel InChI=1S/C13H9ClN2O/c14-8-5-6-11-12(7-8)16-13(17)9-3-1-2-4-10(9)15-11/h1-7,15H,(H,16,17)
SMIL Clc1ccc2Nc3ccccc3C(=O)Nc2c1
IUPAC navn 3-chloro-5,11-dihydrobenzo[b][1,4]benzodiazepin-6-one
Molekylvægt (g/mol) 244.68
Mængde 25 mg
Formel vægt 244.0403 g/mol