Learn More
8-Bromo-6-cyanoimidazo[1,2-a]pyridine, 95%, Alfa Aesar™
Brand: Alfa Aesar H35030.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1221791-93-0 | |
222.045 | |
DWOXNHVFPVGULV-UHFFFAOYSA-N | |
50853181 | |
C1=CN2C=C(C=C(C2=N1)Br)C#N |
C8H4BrN3 | |
MFCD14584821 | |
8-bromoimidazo 1,2-a pyridine-6-carbonitrile, 8-bromo-6-cyanoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-carbonitrile,8-bromo, 8-bromo-4-hydroimidazo 1,2-a pyridine-6-carbonitrile | |
8-bromoimidazo[1,2-a]pyridine-6-carbonitrile |
Specifications
8-Bromo-6-cyanoimidazo[1,2-a]pyridine | |
1g | |
C8H4BrN3 | |
8-bromoimidazo 1,2-a pyridine-6-carbonitrile, 8-bromo-6-cyanoimidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-carbonitrile,8-bromo, 8-bromo-4-hydroimidazo 1,2-a pyridine-6-carbonitrile | |
C1=CN2C=C(C=C(C2=N1)Br)C#N | |
222.045 | |
222.05 |
184°C to 186°C | |
1221791-93-0 | |
MFCD14584821 | |
DWOXNHVFPVGULV-UHFFFAOYSA-N | |
8-bromoimidazo[1,2-a]pyridine-6-carbonitrile | |
50853181 | |
95% |