Få mere at vide
(7S)-7-Amino-5,7-dihydro-5-methyl-6H-dibenz[b,d]azepin-6-one, TRC
Artikelnummer. 30328235
Skift visning
Mængde:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328235 5 mg
30313989 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30328235 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6048955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (7S)-7-Amino-5,7-dihydro-5-methyl-6H-dibenz[b,d]azepin-6-one
CAS 365242-16-6
Molekylær formel C15H14N2O
Mængde 5 mg
InChI formel InChI=1S/C15H14N2O/c1-17-13-9-5-4-7-11(13)10-6-2-3-8-12(10)14(16)15(17)18/h2-9,14H,16H2,1H3/t14-/m0/s1
SMIL CN1C(=O)[C@@H](N)c2ccccc2c3ccccc13
IUPAC navn (7S)-7-amino-5-methyl-7H-benzo[d][1]benzazepin-6-one
Molekylvægt (g/mol) 238.28
Formel vægt 238.1106