Få mere at vide
(7R,8S,9R,10S)-rel-7,8,9,10-Tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol, TRC
Artikelnummer. 30009316
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30009316 0.5 mg 0.5mg
30004599 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009316 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3099950.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61490-66-2
Molekylær formel C20 H16 O4
Molekylvægt (g/mol) 320.34
Synonym Benzo[a]pyrene-7,8,9,10-tetrol, 7,8,9,10-tetrahydro-, (7R,8S,9R,10S)-rel-, Benzo[a]pyrene-7,8,9,10-tetrol, 7,8,9,10-tetrahydro-, (7alpha,8beta,9beta,10alpha)-, rel-(7R,8S,9R,10S)-7,8,9,10-Tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol, (7,10/8,9)-Tetrahydroxy-7,8,9,10-tetrahydrobenzo[a]pyrene, (+/-)-Benzo[a]pyrene-r-7,t-8,t-9,c-10-tetrahydrotetrol, 7beta,8alpha,9alpha,10beta-Tetrahydroxy-7,8,9,10-tetrahydrobenzo[a]pyrene, Benzo[a]pyrenetetrol I 1, Tetrol I 1
IUPAC navn (7R,8S,9R,10S)-7,8,9,10-tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol
SMIL O[C@H]1[C@@H](O)[C@H](O)c2cc3ccc4cccc5ccc(c2[C@@H]1O)c3c45
Tekniske data
Kemisk navn eller materiale (7R,8S,9R,10S)-rel-7,8,9,10-Tetrahydrobenzo[a]pyrene-7,8,9,10-tetrol
Mængde 0.5 mg
InChI formel InChI=1S/C20H16O4/c21-17-13-8-11-5-4-9-2-1-3-10-6-7-12(15(11)14(9)10)16(13)18(22)20(24)19(17)23/h1-8,17-24H/t17-,18+,19+,20-/m1/s1
Formel vægt 320.1049