missing translation for 'onlineSavingsMsg'
Learn More

7Beta-Spironolactone, TRC

Product Code. 30411736
Change view
Click to view available options
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30411736 25 mg
30418646 10 mg
30409511 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411736 Supplier Toronto Research Chemicals Supplier No. TRCS68301025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33784-05-3
Molekylær formel C24 H32 O4 S
Molekylvægt (g/mol) 416.57
Synonym Pregn-4-ene-21-carboxylic acid, 7-(acetylthio)-17-hydroxy-3-oxo-, γ-lactone, (7β,17α)-, 17α-Pregn-4-ene-21-carboxylic acid, 17-hydroxy-7β-mercapto-3-oxo-, γ-lactone, acetate (8CI), 3'-(3-Oxo-7β-acetylthio-17β-hydroxyandrost-4-en-17α-yl)-propionic acid lactone, 7β-(Acetylthio)-17α-hydroxy-3-oxopregn-4-ene-21-carboxylic acid γ-lactone, SC 11940
IUPAC navn S-[(7S,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMIL CC(=O)S[C@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]45CCC(=O)O5)[C@H]13

Specifications

Kemisk navn eller materiale 7-beta-Spironolactone
Mængde 25 mg
InChI formel InChI=1S/C24H32O4S/c1-14(25)29-19-13-15-12-16(26)4-8-22(15,2)17-5-9-23(3)18(21(17)19)6-10-24(23)11-7-20(27)28-24/h12,17-19,21H,4-11,13H2,1-3H3/t17-,18-,19-,21+,22-,23-,24+/m0/s1
Formel vægt 416.2021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.