Få mere at vide
7beta-Hydroxycholesterol-25,26,26,26,27,27,27-d7, CDN
Artikelnummer. 30319604
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319604 0.01 g
30316972 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319604 Leverandør CDN Leverandørnr. CDND41230.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7 beta-Hydroxycholesterol 25,26,26,26,27,27,27-d7
CAS 349553-97-5
Molekylær formel C27 2H7 H39 O2
Mængde 0.01 g
Synonym Cholest-5-ene-25,26,26,26,27,27,27-d7-3,7-diol, (3β,7β)-, (3β,7β)-Cholest-5-ene-25,26,26,26,27,27,27-d7-3,7-diol, 7β-Hydroxycholesterol-25,26,26,26,27,27,27-D7, (3β,7β)-Cholest-5-ene-3,7-diol-d7, 7β-Hydroxycholesterol-d7
InChI formel InChI=1 S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h16-18,20-25,28-29 H,6-15H2,1-5H3/t18-,20+,21-,22+,23+,24+,25+,26+,27-/m1/s1/i1D3,2D3,17 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(CCC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 [C@@H](O)C=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C([2 H])([2 H])[2 H]
IUPAC navn (3 S,7 R,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthrene-3,7-diol
Molekylvægt (g/mol) 409.7
Formel vægt 409.3937 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity