Få mere at vide
7Beta-Hydroxy-4-cholesten-3-one-d7, TRC
Artikelnummer. 30407012
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30407012 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407012 Leverandør Toronto Research Chemicals Leverandørnr. TRCH8251270.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7beta-Hydroxy-4-cholesten-3-one 25,26,26,26,27,27,27-d7
CAS 1262046-74-1
Molekylær formel C27 2H7 H37 O2
Mængde 0.5 mg
Synonym Cholest-4-en-3-one-25,26,26,26,27,27,27-d7, 7-hydroxy-, (7β)-, 7β-Hydroxy-4-cholesten-3-one-d7, 7β-Hydroxy-4-cholesten-3-one-25,26,26,26,27,27,27-D7, (7β)-7-Hydroxycholest-4-en-3-one-25,26,27-d7
InChI formel InChI=1S/C27H44O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h15,17-18,21-25,29H,6-14,16H2,1-5H3/t18-,21-,22+,23+,24+,25+,26+,27-/m1/s1/i1D3,2D3,17D
SMIL [2H]C([2H])([2H])C([2H])(CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC navn (7S,8S,9S,10R,13R,14S,17R)-7-hydroxy-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 407.68
Formel vægt 407.3781