missing translation for 'onlineSavingsMsg'
Få mere at vide

7Alpha-tert-Butyldimethylsilyloxycholest-5-en-3Beta-ol, TRC

Product Code. 30338779
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30338779 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338779 Supplier Toronto Research Chemicals Supplier No. TRCB69121550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 7alpha-tert-Butyldimethylsilyloxycholest-5-en-3beta-ol
CAS 2375033-57-9
Molekylær formel C33H60O2Si
Mængde 50 mg
Synonym (3β,7α)-7-O-tert-Butyldimethylsilyl-cholest-5-ene-3,7-diol
InChI formel InChI=1S/C33H60O2Si/c1-22(2)12-11-13-23(3)26-14-15-27-30-28(17-19-33(26,27)8)32(7)18-16-25(34)20-24(32)21-29(30)35-36(9,10)31(4,5)6/h21-23,25-30,34H,11-20H2,1-10H3/t23?,25-,26+,27-,28-,29?,30-,32?,33?/m0/s1
SMIL O[C@H](C1)CC[C@@]2(C)C1=C[C@@H](O[Si](C)(C)C(C)(C)C)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCCC(C)C
IUPAC navn (3S,7S,8S,9S,10R,13R,14S,17R)-7-((tert-butyldimethylsilyl)oxy)-10,13-dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 516.91
Formel vægt 516.93
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.