Få mere at vide
7Alpha-Hydroxy-4-cholesten-3-one, TRC
Artikelnummer. 30012565
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012565 25 mg
30012011 5 mg
30007973 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012565 Leverandør Toronto Research Chemicals Leverandørnr. TRCH82513025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3862-25-7
Molekylær formel C27 H44 O2
Molekylvægt (g/mol) 400.64
Synonym Cholest-4-en-3-one, 7-hydroxy-, (7α)-, Cholest-4-en-3-one, 7α-hydroxy- (8CI), (7α)-7-Hydroxycholest-4-en-3-one, δ4-7α-Hydroxycholestenone, 7α-Hydroxy-4-cholesten-3-one, 7α-Hydroxycholest-4-en-3-one
IUPAC navn (7R,8S,9S,10R,13R,14S,17R)-7-hydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMIL CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 7a-Hydroxy-4-cholesten-3-one
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C27H44O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h15,17-18,21-25,29H,6-14,16H2,1-5H3/t18-,21-,22+,23+,24-,25+,26+,27-/m1/s1
Formel vægt 400.3341
Procent renhed >95%