Få mere at vide
7Alpha,12Alpha-Dihydroxy-5Beta-cholestan-3-one, TRC
Artikelnummer. 30133212
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133212 10 mg
30008890 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133212 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45241010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 547-97-7
Molekylær formel C27 H46 O3
Molekylvægt (g/mol) 418.65
Synonym (5β,7α,12α)-7,12-Dihydroxy-cholestan-3-one;
IUPAC navn (5R,7R,8R,9S,10S,12S,13R,14S,17R)-7,12-dihydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
SMIL CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4CC(=O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Tekniske data
Kemisk navn eller materiale 7alpha,12alpha-Dihydroxy-5beta-cholestan-3-one
Mængde 10 mg
InChI formel InChI=1S/C27H46O3/c1-16(2)7-6-8-17(3)20-9-10-21-25-22(15-24(30)27(20,21)5)26(4)12-11-19(28)13-18(26)14-23(25)29/h16-18,20-25,29-30H,6-15H2,1-5H3/t17-,18+,20-,21+,22+,23-,24+,25+,26+,27-/m1/s1
Formel vægt 418.3447