Få mere at vide
(7Alpha,11Alpha,12Alpha,17Alpha)-11,12-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic Acid Lactone, Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 11Alpha,12Alpha-Epoxy-7Alpha-(methoxycarbonyl)-3-oxo-17Alpha-pregn-4-ene-21,17-carbolactone |
| CAS | 192704-82-8 |
| Molekylær formel | C24 H30 O6 |
| Mængde | 10 mg |
| Synonym | Pregn-4-ene-7,21-dicarboxylic acid, 11,12-epoxy-17-hydroxy-3-oxo-, 21,17-lactone, 7-methyl ester, (7α,11α,12α,17α)-, Pregn-4-ene-7,21-dicarboxylic acid, 11,12-epoxy-17-hydroxy-3-oxo-, γ-lactone, methyl ester, (7α,11α,12α,17α)- (9CI), Spiro[11,12-epoxy-17H-cyclopenta[a]phenanthrene-17,2'(3'H)-furan], pregn-4-ene-7,21-dicarboxylic acid deriv., 11α,12α-Epoxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21,17-carbolactone, 9α,11α-Deepoxy-11α,12α-epoxyeplerenone, Pregn-4-ene-7,21-dicarboxylic acid, 11,12-epoxy-17-hydroxy-3-oxo-, 21,17-lactone, 7-methyl ester, (7α,11α,12α,17α)-, Pregn-4-ene-7,21-dicarboxylic acid, 11,12-epoxy-17-hydroxy-3-oxo-, γ-lactone, methyl ester, (7α,11α,12α,17α)- (9CI), Spiro[11,12-epoxy-17H-cyclopenta[a]phenanthrene-17,2'(3'H)-furan], pregn-4-ene-7,21-dicarboxylic acid deriv., 11α,12α-Epoxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21,17-carbolactone, 9α,11α-Deepoxy-11α,12α-epoxyeplerenone, (7Alpha,11Alpha,12Alpha,17Alpha)-11,12-Epoxy-17-hydroxy-3-oxo-pregn-4-ene-7,21-dicarboxylic Acid Lactone, Methyl Ester |
| InChI formel | InChI=1S/C24H30O6/c1-22-7-4-13(25)10-12(22)11-14(21(27)28-3)17-15-5-8-24(9-6-16(26)30-24)23(15,2)20-19(29-20)18(17)22/h10,14-15,17-20H,4-9,11H2,1-3H3/t14-,15+,17+,18-,19+,20+,22+,23-,24-/m1/s1 |
| SMIL | COC(=O)[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3[C@@H]4O[C@@H]4[C@@]5(C)[C@@H](CC[C@@]56CCC(=O)O6)[C@H]13 |
| IUPAC navn | methyl (1S,2S,4R,5R,6R,9S,10S,11R,18R)-5,18-dimethyl-5',15-dioxospiro[3-oxapentacyclo[8.8.0.02,4.05,9.013,18]octadec-13-ene-6,2'-oxolane]-11-carboxylate |
| Molekylvægt (g/mol) | 414.491 |
| Formel vægt | 414.204 |