Learn More
Ansell Edmont™ 76-501 Series Blue FiberTuf™ Light Weight Gloves
Product Code. 12696991 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12696991 10 12 pairs
12607001 7 12 pairs
12617001 8 12 pairs
12627001 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12696991 Supplier Ansell Edmont™ Supplier No. 76501100
 Restricted Item
Forventet forsendelse: 15-12-2026
Designed for optimal results in dry applications requiring light mechanical protection, comfort and good grip.
  • Aromatic polyester fiber construction
  • For handling, packaging, light assembly applications
  • Synthetic fiber liner
  • Knitted construction
  • PVC coating
  • Dotted on palm side finish
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material PVC
Certifications/Compliance Standard CATEGORY II EN 388 : 214X
Cuff Material PVC
Accelerator Free Yes
Applications Industrial
Color Blue
Disposable Reusable
Exterior Finish Partially Textured
Latex Free Yes
Material Synthetic fiber
Powdered Powder Free
Product Type Lightweight Gloves
Size 10
Sterility Non-sterile
Glove Cuff Style Knit
Liner Material Polyester Cotton
Palm Type Dotted-palm
Quantity 12 Pairs
Series 76-501