Få mere at vide
SHOWA™ 7580 Single-Use Nitrile Gloves, Powder-Free
Størrelse:
Small
Medium
Large
X-Large
2X-Large
Pakningsstørrelse:
Pakke med 50
Beskrivelse
- Thicker glove provides more resistance to chemicals
- Avoids latex allergies risks type I and type IV
- Second skin feel, softer texture and easy donning
- Low-modulus formulation to improve fit and reduce hand fatigue
- Textured finish on fingertips to enhance grip
- Without plasticisers, powder-free and silicone-free
Tekniske data
Tekniske data
| Certificeringer/overholdelse (Europa) | EU Regulation 425/2016, EN 420, EN 455, EN 374-1:2016, EN 374-5:2016, EU Regulation 1935/2004 & 10/2011 |
| AQL | 0.65 |
| Farve | Cobalt Blue |
| Engangs | Single-use |
| Hånd | Ambidextrous |
| Latex fri | Yes |
| Materiale | Nitrile |
| Pulveriseret | Powder Free |
| Størrelse | Small |
| Sterilitet | Non-sterile |
| Vis mere |