Learn More
7-p-Toluenesulfonylhydrazide 3Beta,4Beta-Dihydroxy-chol-5-ene-24-carboxylic Acid Ethyl Ester, TRC
Product Code. 30453269
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453269 1 mg 1mg
30455807 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30453269 Supplier Toronto Research Chemicals Supplier No. TRCT5359101MG
 Restricted Item
Forventet forsendelse: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-p-Toluenesulfonylhydrazide 3beta,4beta-Dihydroxy-chol-5-ene-24-carboxylic Acid Ethyl Ester
Analyte or Component Names 7-p-Toluenesulfonylhydrazide 3beta,4beta-Dihydroxy-chol-5-ene-24-carboxylic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C34H50N2O6S
InChI Formula InChI=1S/C34H50N2O6S/c1-6-42-30(38)9-7-8-22(3)24-14-15-25-31-26(16-18-33(24,25)4)34(5)19-17-29(37)32(39)27(34)20-28(31)35-36-43(40,41)23-12-10-21(2)11-13-23/h10-13,20,22,24-26,29,31-32,36-37,39H,6-9,14-19H2,1-5H3/b35-28-/t22-,24-,25+,26+,29+,31+,32-,33-,34-/m1/s1
Solution Type Neat
SMILES [H][C@@]1([C@]2(C)C([C@@H](O)[C@@H](O)CC2)=C/3)[C@@]([C@@](CC[C@]4([H])[C@H](C)CCCC(OCC)=O)([H])[C@]4(C)CC1)([H])C3=N/NS(C5=CC=C(C)C=C5)(=O)=O
IUPAC Name ethyl (R)-5-((3S,4R,8S,9S,10R,13R,14S,17R,E)-3,4-dihydroxy-10,13-dimethyl-7-(2-tosylhydrazineylidene)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)hexanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 614.84
Quantity 1 mg
Formula Weight 614.34