missing translation for 'onlineSavingsMsg'
Få mere at vide

7-Oxooctadecanoic Acid Methyl Ester, TRC

Artikelnummer. 30424316
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424316 250 mg
30423949 50 mg
30418527 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30424316 Leverandør Toronto Research Chemicals Leverandørnr. TRCO870160250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2380-22-5
Molekylær formel C19H36O3
Molekylvægt (g/mol) 312.49
Synonym Methyl 7-Ketooctadecanoate, Methyl 7-Oxooctadecanoate
IUPAC navn methyl 7-oxooctadecanoate
SMIL CCCCCCCCCCCC(=O)CCCCCC(=O)OC

Tekniske data

Kemisk navn eller materiale 7-Oxooctadecanoic Acid Methyl Ester
Mængde 250 mg
InChI formel InChI=1S/C19H36O3/c1-3-4-5-6-7-8-9-10-12-15-18(20)16-13-11-14-17-19(21)22-2/h3-17H2,1-2H3
Formel vægt 312.2664
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.