Få mere at vide
7-O-Demethyl Alpha-Narcotine, TRC
Artikelnummer. 30365691
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365691 10 mg
30352826 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30365691 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23128510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68353-55-9
Molekylær formel C21H21NO7
Molekylvægt (g/mol) 399.39
Synonym (3S)-7-Hydroxy-6-methoxy-3-[(5R)-5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-1(3H)-isobenzofuranone, [S-(R*,S*)]-7-hydroxy-6-methoxy-3-(5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl)-1(3H)-isobenzofuranone, 7-Hydroxy-alpha-narcotine
IUPAC navn (3S)-7-hydroxy-6-methoxy-3-[(5R)-4-methoxy-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-3H-2-benzofuran-1-one
SMIL COc1ccc2[C@H](OC(=O)c2c1O)[C@@H]3N(C)CCc4cc5OCOc5c(OC)c34
Tekniske data
Kemisk navn eller materiale 7-O-Demethyl alpha-Narcotine
Mængde 10 mg
InChI formel InChI=1S/C21H21NO7/c1-22-7-6-10-8-13-19(28-9-27-13)20(26-3)14(10)16(22)18-11-4-5-12(25-2)17(23)15(11)21(24)29-18/h4-5,8,16,18,23H,6-7,9H2,1-3H3/t16-,18+/m1/s1
Formel vægt 399.1318