Få mere at vide
7-Nitrocoumarin, TRC
Artikelnummer. 30475832
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475832 50 mg
30465009 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475832 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49584050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19063-58-2
Molekylær formel C9 H5 N O4
Molekylvægt (g/mol) 191.14
IUPAC navn 7-nitrochromen-2-one
SMIL [O-][N+](=O)c1ccc2C=CC(=O)Oc2c1
Tekniske data
Kemisk navn eller materiale 7-Nitrocoumarin
Mængde 50 mg
InChI formel InChI=1S/C9H5NO4/c11-9-4-2-6-1-3-7(10(12)13)5-8(6)14-9/h1-5H
Formel vægt 191.0219