Få mere at vide
7-Methylmemantine Hydrochloride, Mikromol™
Beskrivelse
7-Methylmemantine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 7-Methyl Memantine hydrochloride |
| CAS | 15210-60-3 |
| Analyt- eller komponentnavne | 7-Methyl Memantine hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H23 N . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Tricyclo[3.3.1.13,7]decan-1-amine, 3,5,7-trimethyl-, hydrochloride (1:1), 1-Adamantanamine, 3,5,7-trimethyl-, hydrochloride (8CI), Tricyclo[3.3.1.13,7]decan-1-amine, 3,5,7-trimethyl-, hydrochloride (9CI), 7-Methyl Memantine Hydrochloride, 7-Methylmemantine Hydrochloride, Memantine Related Compound H as Hydrochloride |
| InChI formel | InChI=1S/C13H23N.ClH/c1-10-4-11(2)6-12(3,5-10)9-13(14,7-10)8-11;/h4-9,14H2,1-3H3;1H |
| SMIL | Cl.CC12CC3(C)CC(C)(C1)CC(N)(C3)C2 |
| IUPAC navn | 3,5,7-trimethyladamantan-1-amine;hydrochloride |
| Vis mere |