Få mere at vide
7-Methylguanosine, TRC
Artikelnummer. 30483928
Skift visning
Mængde:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483928 1 g
30467042 100 mg
30469625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483928 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3118351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20244-86-4
Molekylær formel C11H15N5O5
Molekylvægt (g/mol) 297.27
Synonym 2-Amino-6,9-dihydro-7-methyl-6-oxo-9-β-D-ribofuranosyl-1H-purinium, 2-Amino-1,6-dihydro-7-methyl-6-oxo-9-β-D-ribofuranosyl-purinium, N7-Methylguanosine
SMIL [O-]C1=C2[N+](C)=CN([C@H](O3)[C@@H](O)[C@@H](O)[C@H]3CO)C2=NC(N)=N1
Tekniske data
Kemisk navn eller materiale 7-Methylguanosine
Mængde 1 g
InChI formel InChI=1S/C11H15N5O5/c1-15-3-16(8-5(15)9(20)14-11(12)13-8)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19H,2H2,1H3,(H2-,12,13,14,20)/p+1/t4-,6-,7-,10-/m1/s1
Formel vægt 297.1073