missing translation for 'onlineSavingsMsg'
Learn More

7-Methylguanine, TRC

Product Code. 30007227
Change view
Click to view available options
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30007227 1 g
30006764 250 mg
17910905 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007227 Supplier Toronto Research Chemicals Supplier No. TRCM3117901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-76-7
Molekylær formel C6 H7 N5 O
Molekylvægt (g/mol) 165.15
Synonym 2-Amino-1,7-dihydro-7-methyl-6H-purin-6-one, 2-Amino-7-methylhypoxanthine, N7-Methylguanine, NSC 193444, NSC 19647
IUPAC navn 2-amino-7-methyl-1H-purin-6-one
SMIL Cn1cnc2N=C(N)NC(=O)c12

Specifications

Kemisk navn eller materiale 7-Methylguanine
Mængde 1 g
InChI formel InChI=1S/C6H7N5O/c1-11-2-8-4-3(11)5(12)10-6(7)9-4/h2H,1H3,(H3,7,9,10,12)
Formel vægt 165.0651
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.