Få mere at vide
7-Methyl-d3-1-octyl-7,8,8,8-d4 Alcohol, CDN
Artikelnummer. 30316439
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316439 0.05 g -
30319084 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316439 Leverandør CDN Leverandørnr. CDND79450.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Methyloctanol-D7 (isopropyl-D7)
CAS 1794753-02-8
Molekylær formel C9 D7 H13 O
Mængde 0.05 g
Synonym 7-(Methyl-d3)-1-octanol-7,8,8,8-d4, 7-Methyloctanol-d7, 7-Methyloctanol-D7 (isopropyl-D7), 7-(Methyl-d3)-1-octyl-7,8,8,8-d4 alcohol, 7-Methyl-d3-1-octyl-7,8,8,8-d4 Alcohol
InChI formel InChI=1 S/C9H20O/c1-9(2)7-5-3-4-6-8-10/h9-10 H,3-8H2,1-2H3/i1D3,2D3,9 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(CCCCCCO)C([2 H])([2 H])[2 H]
IUPAC navn 7,8,8,8-tetradeuterio-7-(trideuteriomethyl)octan-1-ol
Molekylvægt (g/mol) 151.2976
Formel vægt 151.1954 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity