missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-4-methylquinoline, 98%, Thermo Scientific™

Catalog Number 15435217
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 6238-12-6
Molecular Formula C12H20O
Refractive Index 1.6168
MDL Number MFCD11655579
Quantity 500 mg
Synonym 1-adamantaneethanol, 2-adamantan-1-yl ethanol, 2-1-adamantyl ethanol, 1-admantaneethanol, 1-adamantane ethanol, tricyclo 3.3.1.13,7 decaneethanol 9ci, tricyclo 3.3.1.1 3,7-decane-1-ethanol, 2-tricyclo 3.3.1.1'3,7 dec-1-yl ethanol, 2-3r,5s,7s-adamantan-1-yl ethanol, 2-adamantan-1-ylethanol
InChI Key ZBIDZPHRNBZTLT-UHFFFAOYSA-N
SMILES C1C2CC3CC1CC(C2)(C3)CCO
IUPAC Name 2-(1-adamantyl)ethanol
Molecular Weight (g/mol) 180.29
PubChem CID 110810
Formula Weight 173.21
Percent Purity 98%
Chemical Name or Material 7-Methoxy-4-methylquinoline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'