Få mere at vide
7-Methoxy-1-naphthaleneethanamine Hydrochloride, TRC
Artikelnummer. 30476763
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476763 100 mg
30482572 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30476763 Leverandør Toronto Research Chemicals Leverandørnr. TRCM265585100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 139525-77-2
Molekylær formel C13 H15 N O . Cl H
Molekylvægt (g/mol) 237.73
Synonym 1-Naphthaleneethanamine, 7-methoxy-, hydrochloride (9CI), 2-(7-Methoxy-1-naphthyl)ethanamine hydrochloride, 2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride, Agomelatine Impurity A HCl
IUPAC navn 2-(7-methoxynaphthalen-1-yl)ethanamine;hydrochloride
SMIL Cl.COc1ccc2cccc(CCN)c2c1
Tekniske data
Kemisk navn eller materiale 2-(7-Methoxy-1-naphthyl)ethylamine hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C13H15NO.ClH/c1-15-12-6-5-10-3-2-4-11(7-8-14)13(10)9-12;/h2-6,9H,7-8,14H2,1H3;1H
Formel vægt 237.092