missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxycoumarin, 99%, Thermo Scientific Chemicals

Catalog Number p-144592
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
93-35-6
C9H6O3
162.14
MFCD00006878
ORHBXUUXSCNDEV-UHFFFAOYSA-N
7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
5281426
CHEBI:27510
OC1=CC=C2C=CC(=O)OC2=C1
This item is not returnable. View return policy
93-35-6
C9H6O3
162.14
MFCD00006878
ORHBXUUXSCNDEV-UHFFFAOYSA-N
7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
5281426
CHEBI:27510
OC1=CC=C2C=CC(=O)OC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 228°C to 234°C
Color Beige-Brown to White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 18, V,1, 386
Merck Index 15, 10032
Solubility Information Solubility in water: 1g/100mL (100 c). Other solubilities: freely soluble in acetic acid and dilute alkalies,freely soluble in alcohol and chloroform,sparingly soluble in ether
IUPAC Name 7-hydroxychromen-2-one
Formula Weight 162.14
Percent Purity 99%
Physical Form Powder and Agglomerate
Chemical Name or Material 7-Hydroxycoumarin, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'