Få mere at vide
7ß-Hydroxycholesterol-d7, TRC
Artikelnummer. 30407017
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30407017 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407017 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9179641MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7beta-Hydroxycholesterol 25,26,26,26,27,27,27-d7
CAS 349553-97-5
Molekylær formel C27 2H7 H39 O2
Mængde 1 mg
Synonym Cholest-5-ene-25,26,26,26,27,27,27-d7-3,7-diol, (3β,7β)-, (3β,7β)-Cholest-5-ene-25,26,26,26,27,27,27-d7-3,7-diol, 7β-Hydroxycholesterol-25,26,26,26,27,27,27-D7, (3β,7β)-Cholest-5-ene-3,7-diol-d7, 7β-Hydroxycholesterol-d7
InChI formel InChI=1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h16-18,20-25,28-29H,6-15H2,1-5H3/t18-,20+,21-,22+,23+,24+,25+,26+,27-/m1/s1/i1D3,2D3,17D
SMIL [2H]C([2H])([2H])C([2H])(CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC navn (3S,7R,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-3,7-diol
Molekylvægt (g/mol) 409.7
Formel vægt 409.3937