Få mere at vide
7-Hydroxy Warfarin, TRC
Artikelnummer. 17940575
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17940575 10 mg
17964935 1 mg
17903315 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17940575 Leverandør Toronto Research Chemicals Leverandørnr. TRCH99613010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17834-03-6
Molekylær formel C19 H16 O5
Molekylvægt (g/mol) 324.33
Synonym 2H-1-Benzopyran-2-one, 4,7-dihydroxy-3-(3-oxo-1-phenylbutyl)-, Coumarin, 3-(α-acetonylbenzyl)-4,7-dihydroxy- (8CI), 4,7-Dihydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one, 7-Hydroxywarfarin
IUPAC navn 4,7-dihydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
SMIL CC(=O)CC(C1=C(O)c2ccc(O)cc2OC1=O)c3ccccc3
Tekniske data
Kemisk navn eller materiale 7-Hydroxy Warfarin
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C19H16O5/c1-11(20)9-15(12-5-3-2-4-6-12)17-18(22)14-8-7-13(21)10-16(14)24-19(17)23/h2-8,10,15,21-22H,9H2,1H3
Formel vægt 324.0998
Procent renhed >95%