missing translation for 'onlineSavingsMsg'
Få mere at vide

7-Hydroxy Ondansetron, TRC

Artikelnummer. 30403410
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30403410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30403410 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95115010MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 7-Hydroxy Ondansetron
CAS 126702-17-8
Molekylær formel C18 H19 N3 O2
Mængde 10 mg
Synonym 4H-Carbazol-4-one, 1,2,3,9-tetrahydro-7-hydroxy-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-, 1,2,3,9-Tetrahydro-7-hydroxy-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one, 7-Hydroxyondansetron, GR 63418
InChI formel InChI=1S/C18H19N3O2/c1-11-19-7-8-21(11)10-12-3-6-15-17(18(12)23)14-5-4-13(22)9-16(14)20(15)2/h4-5,7-9,12,22H,3,6,10H2,1-2H3
SMIL Cc1nccn1CC2CCc3c(C2=O)c4ccc(O)cc4n3C
IUPAC navn 7-hydroxy-9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one
Molekylvægt (g/mol) 309.362
Formel vægt 309.148
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.