Få mere at vide
7-Hydroxy Granisetron-d3 Hydrochloride, TRC
Artikelnummer. 30403682
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30403682 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403682 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9426770.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 7-Hydroxygranisetron indazole-1-methyl-d3 hydrochloride
CAS 2124272-03-1
Molekylær formel C18 2H3 H21 N4 O2 . Cl H
Mængde 0.5 mg
Synonym 7-Hydroxy-1-(methyl-d3)-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide hydrochloride, 7-Hydroxygranisetron-d3 hydrochloride, 7-Hydroxygranisetron-D3 hydrochloride (1-(methyl-D3)-indazole), 7-Hydroxy granisetron-d3 hydrochloride
InChI formel InChI=1S/C18H24N4O2.ClH/c1-21-12-5-3-6-13(21)10-11(9-12)19-18(24)16-14-7-4-8-15(23)17(14)22(2)20-16;/h4,7-8,11-13,23H,3,5-6,9-10H2,1-2H3,(H,19,24);1H/t11-,12+,13-;/i2D3;
SMIL Cl.[2H]C([2H])([2H])n1nc(C(=O)N[C@H]2C[C@H]3CCC[C@@H](C2)N3C)c4cccc(O)c14
IUPAC navn 7-hydroxy-N-[(1S,5R)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]-1-(trideuteriomethyl)indazole-3-carboxamide;hydrochloride
Molekylvægt (g/mol) 367.89
Formel vægt 367.1854